Learn More
Simport™ Scientific StainTray™ Lids for Slide Staining System
Catalog No. 22045037 Shop All Simport Scientific Products
Change view
Click to view available options
Material:
ABS
PETE
Color:
Black
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Color
22-045-037 ABS Black
22-045-036 PETE Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-045-037 Supplier Simport™ Scientific Supplier No. M9212
Add to Cart
Edge
Add to Cart
StainTray™ Lids for Slide Staining System for use with SlideTray staining system.
  • A clear PETG lid allowing for visual examination with a temperature range of -20°C to +60°C
  • A black ABS lid for fluorescent work with a temperature range of -80°C to +80°C
  • Lid only
  • For M920
Specifications
Color Black
Material ABS
Type Lid