Learn More
Description
Includes
Each part number represent one side-by-side cabinet; set of three cabinets shown represents three separate items stacked with base.
No interior metal parts to corrode: wooden hinges and handles, polypropylene lining on cabinet floor, and polypropylene padlock assembly.
- Lock with two keys and piano-style door hinges
- All hardware is corrosion resistant
- Liquid-tight trough floors
- 10 x 10 in. (25 x 25cm) Hazard labels on both doors
- Epoxy paint finish inside and out
Corrosives Cabinet
- Capacity: 22 x 2.5L (approx. 14.5 gal./55L)
- Blue finish
- Accepts nitric acid compartment
Flammables Cabinet
- Capacity: 16 x 1 gal. (approx. 60.5L)
- Yellow finish
Combination Cabinet
- Combination corrosives/flammables cabinet
- Corrosives compartment capacity: 10 x 2.5L (approx. 6.5 gal./25L)
- Flammables compartment capacity: 8 x 1 gal. (approx. 30L)
- Gray finish
Floor Stands
- Height: 4.5 in. (11cm)
- Epoxy paint finish
Order Info
Interior: 15.125D x 45W x 16 in.H (40 x 114 x 41cm); Exterior: 18L x 47W x 18 in.H (46 x 119 x 46cm)
Compliance
Meet all applicable OSHA and NFPA standards.
Specifications
Specifications
| Height (Metric) Exterior | 11.43 cm |
| Depth (English) Exterior | 17.5 in. |
| Width (English) Exterior | 46.5 in. |
| For Use With (Equipment) | Combination Acid and flammables Stak-a-Cab cabinet |
| Includes | Four built in adjustable leveling feet |
| Surface Coating | Epoxy |
| Width (Metric) Exterior | 118.11 cm |
| Height (English) Exterior | 4.5 in. |
| Color | Gray |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
