Learn More
Description
- Includes all CBC parameters, reticulocytes and nucleated RBCs.
- Available in three clinically significant levels.
- Plastic vials with pierceable caps for analyzer autosampling.
- Assay values are available for download.
- 14-day open-vial stability, 105-day closed-vial stability.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 10°C |
| Format | Vial |
| For Use With (Equipment) | Beckman Coulter™ UniCel™ DxH™ 800, LH780/785, LH 750/755, LH500 and HmX |
| Type | STaK-Chex™ plus Retics Control |
| Intended Use | For evaluating the accuracy and precision of automated procedures that measure Blood cell parameters |
| Level | 4 Low |
| Quantity | 4 x 4.5 mL |
| Stability | 105 Days |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
