Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ BUN Liquid Reagent for Diagnostic Set
GSA_VASDP

Catalog No. SB0580250 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
250 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB-0580-250 250 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SB-0580-250 Supplier Stanbio™ Supplier No. 0580250

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent liquid for Stanbio™ Diagnostic Set

  • Assay: BUN
  • Assay method: Colorimetric (Diacetylmonoxime)
  • Linear range: 80mg/dL
  • Working stability: 2 years from manufacturing date
  • Kit Contents: Urea Nitrogen Standard, Urea Nitrogen Color Reagent, Urea Nitrogen Acid Reagent

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 15°C to 30°C
Type Urea Nitrogen (BUN) Test Set
For Use With (Application) Diacetylmonoxime methodology
Quantity 250 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.