Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Cholesterol Liquid Reagent for Diagnostic Set
SDP

Catalog No. sb1010430 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
120 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB-1010-430 120 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SB-1010-430 Supplier Stanbio™ Supplier No. 1010430
This item is not returnable. View return policy

Reagent liquid for Stanbio™ Diagnostic Set

  • Assay: Cholesterol
  • Assay method: Colorimetric (CO/POD)
  • Linear range: 750mg/dL
  • Size: 120 Tests
  • Working stability: Until expiration date

Specifications

Content And Storage 2°C to 8°C
Type Cholesterol LiquiColor™ Test (Enzymatic)
For Use With (Application) Total and HDL Cholesterol assays
Quantity 120 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.