Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ CK Liqui-UV™ Test (Creatine Kinase/Rate)
SDP

Catalog No. 22022630 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
4 x 24mL R2, 4 x 6mL R2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-022-630 4 x 24mL R2, 4 x 6mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-022-630 Supplier Stanbio™ Supplier No. 2910430

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use liquid reagents

These liquid-stable testing reagents are designed for ease of use and maximum stability. Reagents are ready to use anytime for routine or Stat analysis with no measure reagent preparation required. A mixture of special additives, collectively called LCF (lipid clearing factor), eliminates turbidity in specimens with increased lipid concentrations, assuring quality results. CK Liqui-UV™ reagents are read in the UV range.

  • Szasz modified reaction
  • NAC activated
  • Working reagent stable up to 3 weeks at 2° to 8°C
  • Linear to 1500U/L
  • Read at 340nm
  • Kit contents: reagents R1 and R2

Specifications

Content And Storage 2°C to 8°C
Type CK Liqui-UV™ Test
Quantity 4 x 24mL R2, 4 x 6mL R2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.