Learn More
Description
Includes
Reagent 1: 4 x 105mL; Reagent 2: 1 x 80 mL; Standard: 1 x 3 mL (100mg/dL)
- Use serum, plasma or urine
- Ready-to-use liquid reagents
- Working reagent stable up to 90 days
- Concentration: Linear to 500mg/dL
- Read at 340nm
Specifications
Specifications
| Type | Glucose Liqui-UV™ Test |
| Includes | R1: 4 x 105mL, R2: 1 x 80mL and standard: 1 x 3mL |
| Quantity | Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
