Learn More
Description
- Assay: Iron TIBC
- Assay method: Colorimetric (Ferrozine)
- Linear range: 500mg/dL
- Size: 110 Tests
- Working stability: Until expiration date
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Type | Iron and Total Iron Binding Capacity (TIBC) Test set |
| For Use With (Application) | Ferrozine methodology |
| Quantity | 110 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
