Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ LiquiColor™ Direct Bilirubin Reagent
SDP

Catalog No. SB0245250 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB-0245-250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SB-0245-250 Supplier Stanbio™ Supplier No. 0245250
This item is not returnable. View return policy

Includes

Reagent: 1 x 250mL, Oxidant: 1 x 15mL

For use with Total Bilirubin test/Modified Walters-Gerarde methodology

  • Volume: 250mL

Specifications

Certifications/Compliance CE Marked
Type Direct Bilirubin Reagent
Includes R: 1 x 250mL, Oxidant: 1 x 15mL
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.