Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Potassium Reagent liquid for Diagnostic Set
SDP

Catalog No. SB0160050 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB-0160-050 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SB-0160-050 Supplier Stanbio™ Supplier No. 0160050

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stanbio™ Reagent liquid for Diagnostic Set

  • Assay: Potassium
  • Assay method: Turbidimetric
  • 10mEq/L
  • 50 Tests
  • Working stability: Until 60 days old

Specifications

Content And Storage 2°C to 8°C
Type Potassium Test Set
For Use With (Application) Turbidimetric methodology
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.