Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Salicylate Liqui-UV™ Test
SDP

Catalog No. SBSR100 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
1 x 50mL R1, 1 x 1mL R2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SBSR100 1 x 50mL R1, 1 x 1mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SBSR100 Supplier Stanbio™ Supplier No. 2410051

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Certifications/Compliance CE Marked
Type Salicylate Test Kit
For Use With (Application) Quantitative testing
Includes 1 x 50mL R1, 1 x 1mL R2, 1 x 3mL (300mg/L) Standard
Quantity 1 x 50mL R1, 1 x 1mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.