Learn More
Description
- For use with Stanbio Sirrus Clinical Analyzers, Cat. Nos. SBG-550-0001, SBG-500-0001
Specifications
Specifications
| Sample Type | Plasma, Serum |
| Type | Urea Nitrogen Liqui-UV™ Test |
| For Use With (Application) | Clinical chemistry, proteins and therapeutic drug monitoring |
| Quantity | 100 Tests |
| Sample Size | 3 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
