Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Sirrus™ General Chemistry Reagents
SDP

Catalog No. SBS2940600 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
100 Tests
125 Tests
Type:
Albumin LiquiColor™ Test
Calcium (Arsenazo) LiquiColor™ Test
Creatinine LiquiColor™ Test
Glucose Liqui-UV™ Test
LDH Liqui-UV™ Test
Total Bilirubin LiquiColor™ Test
Total Protein LiquiColor™ Test
Urea Nitrogen Liqui-UV™ Test
Sample Size:
3 μL
4.5 μL
5 μL
12 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Sample Size
SBS2940600 100 Tests LDH Liqui-UV™ Test 12 μL
SBS0155750 125 Tests Calcium (Arsenazo) LiquiColor™ Test 3 μL
SBS0230600 100 Tests Total Bilirubin LiquiColor™ Test 5 μL
SBS0430600 100 Tests Creatinine LiquiColor™ Test 4.5 μL
SBS0285750 125 Tests Albumin LiquiColor™ Test 3 μL
SBS0250750 125 Tests Total Protein LiquiColor™ Test 3 μL
SBS2020600 100 Tests Urea Nitrogen Liqui-UV™ Test 3 μL
SBS1060600 100 Tests Glucose Liqui-UV™ Test 3 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. SBS2940600 Supplier Stanbio™ Supplier No. S2940600
This item is not returnable. View return policy

  • For use with Stanbio Sirrus Clinical Analyzers, Cat. Nos. SBG-550-0001, SBG-500-0001

Specifications

Sample Type Plasma, Serum
Type LDH Liqui-UV™ Test
For Use With (Application) Clinical chemistry, proteins and therapeutic drug monitoring
Quantity 100 Tests
Sample Size 12 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.