Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™Chemistry Beta-Hydroxybutyrate / TDM Bi-Level Control
GSA_VASDP

Catalog No. SB2465605 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
6 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB2465605 6 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB2465605 Supplier Stanbio™Chemistry Supplier No. 2465605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bi-level (high and low) control kit can provide quality control for any Beta-hydroxybutyrate (BHB) reagent kit. Designed for maximum stability and ease of use

  • The Bi-level Control Solution for Beta-Hydroxybutyrate includes accurate measurements of Acetaminophen, Beta-Hydroxybutyrate, and Salicylate to ensure dependable testing.
  • Ready-to-use liquid controls for immediate use, saving time and effort
  • Compatible with Serum or Plasma: Suitable for use with serum or plasma samples, providing flexibility in sample selection
  • Linear Range up to 4.5 mmol/L: Offers a linear measurement range of up to 4.5 mmol/L for precise results.
  • 60-Day Open Vial Stability: Stable for 60 days after opening when refrigerated (2 to 8°C), minimizing waste.
  • FDA Cleared
  • Matrix-Free Serum Substitute: Utilizes a synthetic serum substitute (SeraSub™) matrix, free of serum infectious agents.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance FDA-cleared
Content And Storage Refrigerated
Detectable Analytes beta-hydroxybutyrate (BHB), Acetaminophen, Salicylate
Type Beta-Hydroxybutyrate / TDM Bi-Level Control
Includes Two levels of unassayed controls with low and high concentrations of analytes including Acetaminophen, Beta-Hydroxybutyrate, and Salicylate.
Level 2
Quantity 6 x 5 mL
Stability 60 days after opening at 2°C to 8°C
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.