Learn More
Stanbio™ Total Protein (CSF/Urine) Liquid Reagent for Diagnostic Set
Shop All Stanbio Laboratory Products

Description
- Assay: Total Protein (CSF/Urine)
- Assay method: Colorimetric
- Linear range: 150mg/dL
- Size: 250mL
- Working stability:1 Year
Specifications
Specifications
| Content And Storage | Room Temperature |
| Type | Protein, Micro LiquiColor™ Test (CSF/Urine) |
| For Use With (Application) | Pyrogallol red/colorimetric methodology |
| Quantity | 250 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.