Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Total Protein Liquid Reagent for Diagnostic Set
SDP

Catalog No. SB0250500 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB-0250-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SB-0250-500 Supplier Stanbio™ Supplier No. 0250500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent liquid for Stanbio™ Diagnostic Set

  • Assay: Total Protein
  • Assay method: Colorimetric (Biuret)
  • 10g/dL
  • 500mL
  • Working stability: Until expiration date

Specifications

Content And Storage Room Temperature
Type Protein, Total LiquiColor™ Test
For Use With (Application) Biuret reaction methodology
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.