Learn More
Sol-M™ Standard 3-Piece Syringe without Needle
Catalog No. 22024272
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
30 mL
60 mL
Connection Type:
Catheter
Fixed
Luer Lock
Slip Tip
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
22-024-272 1 mL Slip Tip
22-024-271 1 mL Luer Lock
22-024-273 1 mL Fixed
22-024-277 3 mL Luer Lock
22-024-278 3 mL Slip Tip
22-024-289 5 mL Luer Lock
22-024-290 5 mL Slip Tip
22-024-292 10 mL Luer Lock
22-024-293 10 mL Slip Tip
22-024-298 20 mL Luer Lock
22-024-299 20 mL Slip Tip
22-024-300 30 mL Luer Lock
22-024-301 60 mL Luer Lock
22-024-302 60 mL Catheter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 22-024-272 Supplier Sol-M™ Supplier No. P180011LDS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to accurately measure liquid medications with low dead space technology that helps reduce medication waste and improve dosing accuracy. Available in a wide range of sizes.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
  • Available in Luer Lock, Luer Slip Tip, and Catheter Tip options
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Disposable Single-use
Packaging Blister
Volume (Metric) 1 mL
Needle Point Style No Needle Included
For Use With (Equipment) Hypodermic Needle, Safety Needle, Blunt Fill Needle
Connection Type Slip Tip
Graduated Yes
Sterility Sterile