Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mopec™ Standard Duty Body Bag with Center Zipper, without Handles

Made from enforced polyehtylene, this standard duty body bag features a center zipper, no handles, and is suitable for cremation.
Supplier: Mopec™ BE176
Description
- Made of Enforced Coextruded Polyethylene
- Straight Center Zipper
- Cremation safe
Specifications
Body Bag | |
Center Zipper | |
Polyethylene | |
8 mil | |
94 in. | |
36 in. |
>200 lb. | |
Black | |
Adult | |
94 x 36 in. | |
238.76 cm | |
91.44 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction