Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO Standard LA Precision Balance

Catalog No. 01804206 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity:
220 g
420 g
2200 g
4200 g
Readability:
1 mg
10 mg
Model:
LA2002E
LA203E
LA4002E
LA403E
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Readability Model
01-804-206 4200 g 10 mg LA4002E
01-804-207 420 g 1 mg LA403E
01-804-205 220 g 1 mg LA203E
01-804-204 2200 g 10 mg LA2002E
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-804-206 Supplier METTLER TOLEDO Supplier No. 30823723
Add to Cart
Add to Cart

METTLER TOLEDO Standard LA Precision Balances are cost efficient, accurate and designed for long lasting performance. These models offer simple weighing and have external adjustment.

Increase efficiency in your daily weighing operations with the bright, backlit LCD display and 3 built-in applications. Robust construction with a metal base and overload protection defend the balance against rough handling and tough weighing environments.

LA Precision Balances include:

  • Adjustment with external weights
  • Bright backlit LCD Display
  • 3 built-in weighing applications
  • Front level indicator
  • Overload protection
  • Metal base, ABS top housing
  • RS232 interface
  • Bluetooth option
  • Transfer weight function for easy data transfer

See how it ranks among our balance lines:

  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
TRUSTED_SUSTAINABILITY

Specifications

Connection Type USB-A, RS232
Capacity 4200 g
Display Type Backlit LCD
Readability 10 mg
Material Aluminum, ABS
Model LA4002E
Stabilization Time 3 sec.
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.