Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Standard Mixture of n-alkanes for System Performance Test, Restek™
SDP

Catalog No. 06802074 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-074 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-074 Supplier Restek Corporation Supplier No. 31633
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Reference mixtures for ISO/DIS 9377 (German H-53), gas chromatography; flame ionization detection (GC-FID) method
  • For GC analysis of total petroleum hydrocarbons (TPH) in water
  • Environmentally safer than previous methods
  • Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil.
Compounds:
  • C10) n-Decane (124-18-5)
  • (C12) n-Dodecane (112-40-3)
  • (C14) n-Tetradecane (629-59-4)
  • (C16) n-Hexadecane (544-76-3)
  • (C18) n-Octadecane (593-45-3)
  • (C20) n-Eicosane (112-95-8)
  • (C22) n-Docosane (629-97-0)
  • (C24) n-Tetracosane (646-31-1)
  • (C26) n-Hexacosane (630-01-3)
  • (C28) n-Octacosane (630-02-4)
  • (C30) n-Triacontane (638-68-6)
  • (C32) n-Dotriacontane (544-85-4)
  • (C34) n-Tetratriacontane (14167-59-0)
  • (C36) n-Hexatriacontane (630-06-8)
  • (C38) n-Octatriacontane (7194-85-6)
  • (C40) n-Tetracontane (4181-95-7)

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Packaging Ampule
CAS Max % 99.920000
Ignition Point 245°C
Odor Pungent
Vapor Density 2.9 (Air = 1)
Flash Point -4°C
Specific Gravity 0.779 -0.784 g/cm3 at 20°C
Concentration 50 μg/mL each in cyclohexane
UN Number UN1145
Color Colorless
Melting Point 6.5°C
Solubility Negligible
Type Multi-component Standard
Quantity 1 mL
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category Danger
Hazard Statement Highly flammable liquid and vapour
May be fatal if swallowed and enters airways
Causes skin irritation. May cause drowsiness or dizziness
Very toxic to aquatic life
Very toxic to aquatic life with long lasting effects
Precautionary Statement Keep away from heat/sparks/open flames/hot surfaces
No smoking. Ground/bond container and receiving equipment
Use explosion proof electrical/ventilation and lighting equipment
Use only non sparking tools
Take precautionary measures against static discharge
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wash hands and skin thoroughly after handling.
Use only outdoors or in a well  ventilated area
Avoid release to the environment
Wear protective gloves/protective clothing/eye protection/face protection
ShelfLife 6 months
EINECSNumber 203-806-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.