Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Standard Mixture of n-Paraffins (consists of C17, C18, C19 and C20) [Standard Material for GC], TCI America™
SDP

Catalog No. S04565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04565G Supplier TCI America Supplier No. S04565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00197833
Synonym n-Paraffin Mixture

Specifications

Physical Form Crystal/Lump
Color White
Quantity 5 g
Chemical Name or Material Standard Mixture of n-Paraffins (consists of C17, C18, C19 and C20) [Standard Material for GC]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.