Learn More
Electron Microscopy Sciences Particle Size Standard Nanosphere 900nm 15 ML

Supplier: Electron Microscopy Sciences 70893

Polystyrene sphere particle-size standards are certified for mean diameter and are traceable to the National Bureau of Standards (NBS). Use for calibration of electron microscopes for viruses, bacteria, and sub-cellular. Packaged as aqueous suspensions.
Certified Mean diameter: 900nm
Nominal Mean diameter: 895nm+/-8.0nm
Size Uniformity std. dev. & C.V: 9.1 nm (1.0%)
Solids content: 1%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.