Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Standard Organic Chemistry Glassware Kit, 9 Piece
SDPSCI_ED

Catalog No. S15697 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
9 Pieces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S15697 9 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15697 Supplier United Scientific™ Supplier No. ORGKIT09
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economic option for organic chemistry work.

  • This starter set is designed for organic chemistry labs and is suitable for small-scale procedures up to 30 g. All of the ground glass components in this set have a joint size of 14/23. Includes a sturdy storage box with foam insert.
  • Contains one each of:
    • Dropping Funnel (50 mL with Rotoflo Stopcock)
    • Receiver Delivery Adapter, Long Stem
    • Stopper for Dropping Funnel
    • Screw Cap Adapter
    • Air Leak / Steam Inlet Tube
    • Thermometer
    • Boiling Flask (50 mL)
    • Still Head
    • Liebig Condenser

Specifications

Material Glass
Quantity 9 Pieces
Product Type Organic Chemistry Kit

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.