Learn More
ABS Standard Pharmacy/Vaccine Refrigerator
GREENER_CHOICESDP
Catalog No. 22651518 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
Capacity (English):
16 cu. ft.
47 cu. ft.
72 cu. ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22-651-518 72 cu. ft.
22-651-506 16 cu. ft.
22-651-514 47 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-651-518 Supplier ABS Supplier No. PHABTHCS72G
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
These ABS pharmacy refrigerators deliver professional results for medical, clinical, and vaccine cold storage.
Experience a range of innovative features to enhance your work.
  • Microprocessor temperature control
  • Array of temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
Our Standard line provides multi-functional benefits in a cost-effective design. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Specifications
Type Refrigerator
Capacity (English) 72 cu. ft.
Capacity (Metric) 2039 L
Temperature Range 2°C to 8°C
Door Style Glass Swing
No. of Doors 3
Shelves 12
Cabinet Material Scratch-resistant Painted Steel
Exterior Finish Powder-coated Steel
Certifications/Compliance ETL Listed
Style Upright
Monitoring Options Alarms, Remote Alarm Contacts
Compressor H-drive
Plug Type NEMA 5-15
Voltage 115 V
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov