Learn More
Stoelting™ Standard Probe Holders
SDP
Catalog No. 10000158 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Electrode Holder, C Clamp
Replacement C Clamp
Replacement Corner Clamp
Replacement Plastic Block
Replacement Side Clamp
Standard Probe Holder - Side Clamp
Standard Probe Holder-Corner Clamp
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-158 Standard Probe Holder - Side Clamp
10-000-157 Standard Probe Holder-Corner Clamp
10-000-160 Electrode Holder, C Clamp
10-000-165 Replacement Corner Clamp
10-000-166 Replacement Side Clamp
10-000-167 Replacement Plastic Block
10-000-168 Replacement C Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-000-158 Supplier Stoelting™ Supplier No. 51632
Add to Cart
Add to Cart
Stoelting Standard Probe Holders include an 11 mm square acrylic block connected to a standard 19.5 cm standard shaft (6 mm diameter) with grooves spaced 1 mm apart. The grooves (on three sides) facilitate loading of probes.
  • Corner Clamp Probe Holder holds shafts from 0.2 to 2.0 mm
  • Side Clamp holds probes from 0.2 to 4.5 mm
  • C Clamp holds probes from 0.2 to 4.5 mm and is useful to release implanted probes
  • Standard Probe Holder-Corner Clamp is included with each Lab Standard™ and has an 19.5 cm shaft with 11 mm square acrylic block
Specifications
For Use With (Application) Neuroscience
Model Standard Probe Holder - Side Clamp