Description
Stoelting Standard Probe Holders include an 11 mm square acrylic block connected to a standard 19.5 cm standard shaft (6 mm diameter) with grooves spaced 1 mm apart. The grooves (on three sides) facilitate loading of probes.
- Corner Clamp Probe Holder holds shafts from 0.2 to 2.0 mm
- Side Clamp holds probes from 0.2 to 4.5 mm
- C Clamp holds probes from 0.2 to 4.5 mm and is useful to release implanted probes
- Standard Probe Holder-Corner Clamp is included with each Lab Standard™ and has an 19.5 cm shaft with 11 mm square acrylic block
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Model | Replacement Corner Clamp |