Learn More
Description
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes
- Membrane grade
- Membrane type: RC
- Molecular weight cut-off: 12 to 14K
Specifications
Specifications
| Product Type | Spectra/Por 4 Dialysis Tubing |
| MWCO | 12 to 14 kDa |
| Material | Regenerated Cellulose |
| Diameter (Metric) | 6.4 mm |
| Diameter (English) | 0.25 in. |
| Length (English) | 100 ft. |
| Length (Metric) | 30 m |
| Formulation | Contain glycerin as a humectant |
| Temperature Range (English) | 39.2°F to 249.8°F |
| Temperature Range (Metric) | 4°C to 121°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
