Learn More
Ampco Safety Tools Standard Socket, 3/8 in. Drive (Metric)
SDP
Catalog No. 19174588 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
13 mm
14 mm
15 mm
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-174-588 18 mm
19-174-580 10 mm
19-174-581 11 mm
19-174-582 12 mm
19-174-583 13 mm
19-174-584 14 mm
19-174-585 15 mm
19-174-586 16 mm
19-174-587 17 mm
19-174-589 19 mm
19-174-590 20 mm
19-174-591 21 mm
19-174-592 22 mm
19-174-593 23 mm
19-174-594 24 mm
19-174-595 25 mm
19-174-596 7 mm
19-174-597 8 mm
19-174-598 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-588 Supplier Ampco Safety Tools Supplier No. SS3/8D18MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Standard Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Standard Socket Wrench
Drive Size 3/8 in.
Length (Metric) 18 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.