Learn More
Leica Biosystems Standard Specimen Clamp 50 x 55 mm for Leica HistoCore Microtomes
Supplier:  Leica Biosystems 14050238005
Encompass
The Leica Biosystems standard specimen clamp (50 x 55 mm) secures rectangular tissue blocks during microtomy and serves as a direct-replacement clamp for compatible HistoCore microtomes. Supplied as a single silver-colored unit, it provides a stable hold for routine sectioning. Confirm instrument fitment before ordering.
  • Holds rectangular tissue blocks measuring up to 50 x 55 mm.
  • Provides stable clamping for consistent sectioning.
  • Supplied as a single unit for straightforward replacement.
  • Silver-colored finish for easy visual identification.
  • Material not specified; follow instrument instructions for use.
Catalog No. NC2753455