Learn More
Electron Microscopy Sciences Gilder Standard Square 50 mesh Copper Grid

Supplier: Electron Microscopy Sciences G50CU

A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Copper
Pitch: 500 µm
Hole: 420 µm
Bar: 80 µm
:
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.