Description
- Stainless steel
- Corrosion resistant
- Strong spring closed forced jaw designed for securing standard taper joints
Specifications
Specifications
| Quantity | 10/Pk. |
| Type | Standard Taper Clamp |
| Material | Stainless Steel (316Ti) |
| For Use With (Equipment) | Standard taper joints |
| Size | 19 |