Learn More
Bal Supply Standard Taper Clamps
Catalog No. 13684145 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Size:
14
19
24
29
45
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
13-684-145 29
13-684-142 14
13-684-143 19
13-684-144 24
13-684-146 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13-684-145 Supplier Bal Supply Supplier No. 203029
Add to Cart
Add to Cart
Stainless Steel 316Ti
  • Stainless steel
  • Corrosion resistant
  • Strong spring closed forced jaw designed for securing standard taper joints
Specifications
Quantity 10/Pk.
Type Standard Taper Clamp
Material Stainless Steel (316Ti)
For Use With (Equipment) Standard taper joints
Size 29