Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keck Standard Taper Clip Assortment

Catalog No. 15700154 Shop All Oakton Products
Change view
Click to view available options
Material:
POM
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
15-700-154 POM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-700-154 Supplier Keck Supplier No. 0684050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economical, Standard Taper Clips

  • Clips securely hold conical joints under pressure without damaging glass
  • Made from polyoxymethylene (POM) acetal resin
  • Resistant to concentrated alkalis and dilute acids
  • Not resistant to halogen gases and concentrated acids
  • Maximum Temperature: 302°F (150°C)

Specifications

Quantity 27/Pk.
Type Standard Taper Clip Assortment
Material POM
For Use With (Application) Securely holding conical joints under pressure
Temperature (Metric) Resistant 150°C
Temperature (English) Resistant 302°F
Color Assorted
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.