Learn More
Description
- Nickel Plated Steel
- Can withstand temperatures up to 932°F
- For Standard Taper Joints
Specifications
Specifications
| Quantity | 10/Pk. |
| Type | Standard Taper Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | Standard Taper Joints |
| Size | 19 |
| Color | Silver |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.