Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Standard Trauma Kit

Catalog No. 19090692 Shop All Medique Products
Change view
Click to view available options
Dimensions (L x W x H):
17 x 9 x 8 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
19-090-692 17 x 9 x 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-692 Supplier Medique Supplier No. 73901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For first responders to handle emergencies.

  • Contents in sturdy Cordura bags with easy to carry shoulder straps
  • Contains mix of items to handle minor emergencies

Specifications

For Use With (Application) Handle Minor Emergencies
Product Type Filled Trauma Kit
Includes Mix of items to handle emergencies, Sturdy Cordura bag with shoulder straps
Size Standard
Sufficient For First Responders
Dimensions (L x W x H) 17 x 9 x 8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.