Learn More
PIP™ Standard Weight 20 GSM SBP Pant with Elastic Waist
Catalog No. 17369783 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-369-783 Medium
17-369-784 Large
17-369-786 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-369-783 Supplier PIP™ Supplier No. 3516/M
Only null left
Add to Cart
Add to Cart
Spunbound polypropylene offers lightweight protection against dry particulates, keeping dirt and debris off the body. Good for use in general labor, pharmaceuticals, cleaning and food processing.
  • Elastic waistband for comfort.
  • Wide leg opening for easy donning.
  • Designed for a roomy fit.
  • High level of comfort and breathability
  • Perfect when full protection is not needed.
Specifications
Size Medium
Product Type Single-use Pants
Gender Unisex
Material Spunbond Polypropylene
Color White
Ankle Style Open
Seam Type Serged
No. of Pockets None
Disposable Single-use
Latex Free No
Sterility Non-sterile