Learn More
Mopec Standing Height, Base Cabinet, 3 Drawers
SDP
Catalog No. 21381167 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Base Cabinet
Catalog No. Product Type
21-381-167 Base Cabinet
1 options
Catalog No. 21-381-167 Supplier Mopec Supplier No. LC12515S
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • Standing height base cabinet with 3 drawers, (1) drawer 7.4 in. H. (1) drawer 9.9 in. H, and (1) drawer 12.4 in. H.
  • Standing Height Cabinets are 35.125 in. tall with the leveling bolt zeroed out and 22 in. deep.
Materials
  • 304 Stainless Steel Construction with a polished No. 4 Finish
Dimensions
  • LC125-15S: 15.0 in. L x 22.0 in. W x 35.1 in. H
Specifications
Product Type Base Cabinet
Color Light Gray
Material Stainless Steel