Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Standing Height Cabinet, LH Door and Slam Latch
SDP

Catalog No. 21381182 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Standing Height Cabinet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
21-381-182 Standing Height Cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-381-182 Supplier Mopec Supplier No. LC18324SLAS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.

Construction

  • A 2 in. deep liquid-tight pan covers the entire bottom of the cabinet to contain liquid leaks and spills. Two diametrically opposed vents with spark screens are provided in the back of the cabinet as well as a grounding screw.
  • Includes a door that is self-closing and is equipped with a three-point latching system that automatically engages when the door closes.
  • Each door is equipped with a fusible link hold-open feature that ensures the door closes should the temperature outside the cabinet exceed 165°F.

Materials

  • 304 stainless steel construction with a polished No. 4 finish

Dimensions

  • LC183-24LAS 24.0 in. L x 22.0 in. W x 35.1 in. H

Performance

  • Cabinets are labeled: FLAMMABLE—KEEP FIRE AWAY
  • Solvent Storage Cabinets are specifically designed for the storage of flammable and combustible liquids

Specifications

Product Type Standing Height Cabinet
Color Light Gray
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.