Learn More
Mopec Standing Height, Corner Door, Base Cabinet
SDP
Catalog No. 21403135 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Corner/Sink Cabinet
Catalog No. Product Type
21-403-135 Corner/Sink Cabinet
1 options
Catalog No. 21-403-135 Supplier Mopec Supplier No. LC19032SE
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • All door cabinets have 1 adjustable shelf.
  • Doors are hinged on the right-hand side
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • 32 in. L x 22 in. W on one side, 32 in. W on the other x 35.1 in. H
Specifications
Product Type Corner/Sink Cabinet
Color Light Gray
Material Stainless Steel