Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride, Dihydrate, Crystal, Reagent, ACS, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18606985 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-985 25 g Poly Bottle
18-606-986 125 g Poly Bottle
18-606-988 500 g Poly Bottle
18-606-987 6 x 125 g Poly Bottle
18-606-990 2.5 kg Poly Pail
18-606-989 6 x 500 g Poly Bottle
18-606-991 4 x 2.5 kg Poly Pail
18-606-992 12 kg Poly Pail
18-606-993 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-606-985 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S153525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1535, 10025-69-1, SnCl2·2H2O

Spectrum™ Chemical Stannous Chloride, Dihydrate, Crystal, Reagent, ACS is also known as Tin Chloride, has a white solid crystal like aspect. Mostly used in electrolytic baths destined for tin-plating, it is also widely used as a reducing agent. In the textile industry it is employed as a mordant to increase the brightness of certain colors in the fabric. It is also employed by metal enthusiasts to determine platinum and gold group metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
IUPAC Name λ²-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98 to 103%
Grade ACS
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.