Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride Dihydrate, MP Biomedicals

Catalog No. ICN10294990 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10294990 500 g
ICN10294991 1 kg
ICN10294980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10294990 Supplier MP Biomedicals, Inc Supplier No. 0210294990
Add to Cart
Add to Cart

Cell Culture Reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2 · 2H2O

Specifications

Quantity 500 g
Formula Weight 225.6
Chemical Name or Material Stannous Chloride Dihydrate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.