Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride Dihydrate, Purified (Tin (II) Chloride Dihydrate), Reagents

Catalog No. 01336946
Change view
Click to view available options
Quantity:
100 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-946 2.5 kg
01-336-947 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-946 Supplier Reagents Holdings Llc Supplier No. C23300002.5A5
Add to Cart
Add to Cart

Stannous Chloride Dihydrate, Purified (Tin (II) Chloride Dihydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
Synonym Tin(II) Chloride Dihydrate
IUPAC Name λ²-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Color White to Off-White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.71
Chemical Name or Material Stannous Chloride Dihydrate, Purified (Tin (II) Chloride Dihydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.