Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stannous Chloride, For Arsenic, 40%, LabChem™

Catalog No. LC252007 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC252007 125 mL
LC252001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC252007 Supplier LabChem, Inc. Supplier No. LC252007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-01-0 , 10025-69-1
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
CAS Max % 60, 40
Linear Formula SnCl2·2H2O
UN Number UN1789
Solubility Information Soluble in water
Formula Weight 225.65
Percent Purity 40%
Identification Passes Test
Packaging Amber Glass
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Stannous Chloride, For Arsenic
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection, protective clothing, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center or doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center or doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center or doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center or doctor.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.