Learn More
3M™ Petrifilm™ Staph Express Disk, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S30735 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
100 Pack
20 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30735 20 Pack
S30711 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30735 Supplier 3M™ Petrifilm™ Supplier No. 6492
Add to Cart
Add to Cart
Used to identify Staphylococcus aureus from all suspect colonies for samples with high background flora
  • Utilizes convenience and efficiency of the 3M™ Petrifilm™ Plate platform
  • Proven cost effective tool for detection of Staph. aureus
  • Results in as few as 22 hours
Specifications
Detects Staphylococcus aureus
Product Type Disk
Test Type Animal Feed, Pet Food
For Use With (Application) Identifying Staphylococcus aureus from suspect colonies with high background flora
Quantity 20 Pack