Learn More
Thermo Scientific™ Staphaurex™ Plus Latex Agglutination Test
Identify staphylococci which possess clumping factor, protein A, and/or surface antigens characteristic of MRSA and MSSA strains.
$614.50 - $1188.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | Staphaurex Plus Latex Agglutination Test |
CE Marker | Yes |
Detectable Analytes | Staphylococcus species |
Type | Staphylococcus aureus Test Kit |
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
R30950102
|
Thermo Scientific™
R30950102 |
Staphaurex Plus Latex Agglutination Test | 150 Tests/Kit | 2°C to 8°C |
Pack of 150 for $614.50
|
|
|||
R30950201
|
Thermo Scientific™
R30950201 |
Staphaurex Plus Latex Agglutination Test | 450 Tests/Kit | 2°C to 8°C |
Pack of 450 for $1,188.00
|
|
|||
Description
Identify staphylococci which possess clumping factor, protein A, and/or surface antigens, including those characteristic of Methicillin Resistant Staphylococcus aureus (MRSA) and Methicillin Sensitive Staphylococcus aureus (MSSA) strains, using the Thermo Scientific™ Staphaurex™ Plus Latex Agglutination Test.
Use this rapid latex agglutination test which utilises yellow latex particles coated with human fibrinogen for the detection of clumping factor, specific IgG for the detection of protein A, and surface antigens characteristic of MRSA and MSSA strains.In the presence of target organisms, the latex particles agglutinate to give a strong and easily visible positive reaction. In the absence of target organisms the particles remain in smooth suspension.
Proven Performance
- Rely on tried-and-true tests with more than 50 years of combined proven performance
- Maintain confidence with greater than 99% sensitivity (according to data available in the package inserts)
Workflow Efficiency
- Clear-cut, robust reactions provide easy-to-read results
- Improve productivity with compact kit configurations, color-coded reagents, and convenient workstations
Economical Solution
- Maximize spending efficiency with the option of purchasing separate kit components
- Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Yes | |
Staphylococcus aureus Test Kit | |
Staphylococcus aureus |
Staphaurex Plus Latex Agglutination Test | |
Staphylococcus species | |
Test latex, disposable reaction cards, disposable sampling and mixing sticks |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.