Learn More
Description
- Biosafety Level: 2
- 2 or 6 self-contained units of a single organismmecA positive; mupA positive; CLSI screening control for high level mupirocin resistance mediated by the mupA gene; SCCmec type II positive; Methicillin resistant
Specifications
Specifications
| ATCC® | ATCC BAA-1708 |
| Biosafety Level | 2 |
| Certifications/Compliance | Clinical Laboratory Standards Institute (CLSI) |
| Content And Storage | 2°C to 8°C |
| Format | KWIK-STIK |
| Manufacturing Origin | United States |
| Product Type | Quality Control Organism |
| For Use With (Application) | Antimicrobial Susceptibility Testing, Microbial Identification |
| Packaging Type | Foil Sleeves |
| Quantity | 2 Pack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
