Learn More
Invitrogen™ Staphylococcus aureus Exfoliative toxin A Polyclonal Antibody

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | Staphylococcus aureus Exfoliative toxin A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | Epidermolytic toxin A; eta |
| Host Species | Rabbit |
| Immunogen | Recombinant Staphylococcus aureus Exfoliative toxin A protein (39-280aa). |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.