Learn More
Invitrogen™ Staphylococcus aureus Monoclonal Antibody (Staph11-232.3), Invitrogen™
Shop All Thermo Scientific ProductsDescription
MA1-10708 detects Staphylococcus aureus from Staph. Aureus and Staph. epidermidis samples. This antibody does not cross-react with Streptococcus sp., Mycoplasma sp. or Corynebacteria sp. MA1-10708 has been successfully used in ELISA applications. The MA1-10708 immunogen is UV-inactivated Staphylococcus aureus cells (ATCC #29740).
Specifications
Specifications
| Antigen | Staphylococcus aureus |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | Staph11-232.3 |
| Concentration | 1.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2-7.4 |
| Gene Alias | MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant |
| Host Species | Mouse |
| Immunogen | UV-inactivated Staphylococcus aureus cells. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.