Learn More
Description
The PA1-73172 antibody reacts with Staphylococcus aureus. PA1-73172 has been successfully used in Immunofluorescence applications. Acetone fixation of the antigen source is recommended prior to staining. The PA1-73172 antibody was raised against soluble and structural antigens of the whole bacterium; ATCC #27660. Recommended short term of less of 6 months store at 2-8°C in the dark. For longer term storage, store at -20°C. Store in dark.
Specifications
Specifications
| Antigen | Staphylococcus aureus |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant |
| Host Species | Rabbit |
| Immunogen | Staphylococcus aureus whole cells. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
