Learn More
ATCC Staphylococcus epidermidis Strain 4477-2 (ATCC 700565), Freeze-dried BSL-2 Clinical Isolate for Antimicrobial Research
Supplier:  ATCC 700565
Encompass
Staphylococcus epidermidis strain 4477-2 (ATCC 700565) is a genome-sequenced, human-derived clinical isolate supplied as a freeze-dried culture for antimicrobial resistance and drug development research. This BBL-2 organism has a defined susceptibility profile and characteristic colony morphology on trypticase soy medium.
  • Genome sequenced.
  • Provided as a freeze-dried viable culture.
  • Biosafety level: BSL 2.
  • Recommended medium: trypticase soy agar/broth (medium 18).
  • Incubation temperature: 37 °C under aerobic conditions.
  • Human blood isolate from Richmond, Virginia (1997).
  • Characteristic colony morphology on medium 18: entire, glistening, circular, smooth, opaque, and low convex.
  • Susceptibility profile: resistant to multiple beta-lactams and other agents; susceptible to gentamicin, tetracycline, and vancomycin.
Catalog No. NC3729161